Anti-MKRN1 Mouse Monoclonal Antibody [clone: 4E9]
Catalog # ABNOH00023608-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Makorin Ring Finger Protein 1
- Antigen symbol:MKRN1
- Clonality:Monoclonal
- Clone:4E9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:23608
- Antigen synonyms:RNF61
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: MKRN1
Clonality: Monoclonal
Clone: 4E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human