Order Entry
Poland
ContactUsLinkComponent
Anti-MORC3 Mouse Polyclonal Antibody
Anti-MORC3 Mouse Polyclonal Antibody
Catalog # ABNOH00023515-B01P
Supplier:  Abnova
undefined
Anti-MORC3 Mouse Polyclonal Antibody
Catalog # ABNOH00023515-B01P
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    MORC Family CW-Type Zinc Finger 3
  • Antigen symbol:
    MORC3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    23515
  • Antigen synonyms:
    MORC family CW-type zinc finger protein 3|Zinc finger CW-type coiled-coil domain protein 3|MORC family CW-type zinc finger protein 3,KIAA0136|ZCWCC3
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pk:
    50 µg

Specifications

About this item

Type: Primary
Antigen: MORC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human