Anti-DUSP12 Mouse Polyclonal Antibody
ABNOH00011266-A01
: Abnova
- Antibody type:Primary
- Antigen name:Dual specificity phosphatase 12
- Antigen symbol:DUSP12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:11266
- Antigen synonyms:YVH1|serine/threonine specific protein phosphatase|dual specificity protein phosphatase 12|DUSP1|dual specificity tyrosine phosphatase YVH1|YVH1 protein-tyrosine phosphatase ortholog
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: DUSP12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human