Order Entry
Poland
ContactUsLinkComponent
Anti-UBE2C Mouse Monoclonal Antibody [clone: 9D3]
Anti-UBE2C Mouse Monoclonal Antibody [clone: 9D3]
Catalog # ABNOH00011065-M01
Supplier:  Abnova
undefined
Anti-UBE2C Mouse Monoclonal Antibody [clone: 9D3]
Catalog # ABNOH00011065-M01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Ubiquitin-conjugating enzyme E2C
  • Antigen symbol:
    UBE2C
  • Clonality:
    Monoclonal
  • Clone:
    9D3
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    11065
  • Antigen synonyms:
    UbcH10|Ubiquitin carrier protein C|Ubiquitin-protein ligase C|Ubiquitin-conjugating enzyme E2 C
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: UBE2C
Clonality: Monoclonal
Clone: 9D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human