Order Entry
Poland
ContactUsLinkComponent
Anti-CPSF6 Mouse Monoclonal Antibody [clone: 1C5]
Anti-CPSF6 Mouse Monoclonal Antibody [clone: 1C5]
Catalog # ABNOH00011052-M07
Supplier:  Abnova
undefined
Anti-CPSF6 Mouse Monoclonal Antibody [clone: 1C5]
Catalog # ABNOH00011052-M07
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    cleavage And polyadenylation specific factor 6, 68kDa
  • Antigen symbol:
    CPSF6
  • Clonality:
    Monoclonal
  • Clone:
    1C5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    11052
  • Antigen synonyms:
    CFIM|HPBRII-7|HPBRII-4|CFIM68
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: CPSF6
Clonality: Monoclonal
Clone: 1C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse