Anti-SLC35D2 Mouse Monoclonal Antibody [clone: 1H5]
Catalog # ABNOH00011046-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Solute Carrier Family 35 Member D2
- Antigen symbol:SLC35D2
- Clonality:Monoclonal
- Clone:1H5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:11046
- Antigen synonyms:UGTrel8|hfrc|HFRC1|SQV7L|HFRC|Homolog of Fringe connection protein 1|SLC35D2|UGTREL8
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: SLC35D2
Clonality: Monoclonal
Clone: 1H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human