Anti-RAB40B Mouse Monoclonal Antibody [clone: 1A1]
Catalog # ABNOH00010966-M01A
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ras-related protein Rab-40B
- Antigen symbol:RAB40B
- Clonality:Monoclonal
- Clone:1A1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:10966
- Antigen synonyms:SEC4L|RAR
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: RAB40B
Clonality: Monoclonal
Clone: 1A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human