Anti-CAPN9 Mouse Monoclonal Antibody [clone: 3A6]
ABNOH00010753-M02
: Abnova
- Antibody type:Primary
- Antigen name:Calpain 9
- Antigen symbol:CAPN9
- Clonality:Monoclonal
- Clone:3A6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:10753
- Antigen synonyms:nCL-4|Calpain-9|New calpain 4|NCL4|CAPN9|Digestive tract-specific calpain|Protein CG36
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: CAPN9
Clonality: Monoclonal
Clone: 3A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human