Anti-MTHFS Mouse Monoclonal Antibody [clone: 2C12]
Catalog # ABNOH00010588-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:5,10-methenyltetrahydrofolate synthetase (5-formyltetrahydrofolate cyclo-ligase)
- Antigen symbol:MTHFS
- Clonality:Monoclonal
- Clone:2C12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:10588
- Antigen synonyms:MTHFS |5,10-methenyltetrahydrofolate synthetase (5-formyltetrahydrofolate cyclo-ligase) |HsT19268|FLJ30410
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: MTHFS
Clonality: Monoclonal
Clone: 2C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human