Anti-PIBF1 Mouse Polyclonal Antibody
ABNOH00010464-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Progesterone-induced-blocking factor 1
- Antigen symbol:PIBF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:10464
- Antigen synonyms:CEP90|C13orf24|progesterone immunomodulatory binding factor 1|PIBF
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: PIBF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human