Anti-TAB1 Mouse Monoclonal Antibody [clone: 2G9]
Catalog # ABNOH00010454-M01A
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:TGF-beta activated kinase 1
- Antigen symbol:TAB1
- Clonality:Monoclonal
- Clone:2G9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:10454
- Antigen synonyms:TAB1|Mitogen-activated protein kinase kinase kinase 7-interacting protein 1|TGF-beta-activated kinase 1 and MAP3K7-binding protein 1|MAP3K7IP1|TAK1-binding protein 1|TGF-beta-activated kinase 1-binding protein 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: TAB1
Clonality: Monoclonal
Clone: 2G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human