Anti-TESK2 Mouse Monoclonal Antibody [clone: 5G1]
ABNOH00010420-M08
: Abnova
- Antibody type:Primary
- Antigen name:Testis-specific kinase 2
- Antigen symbol:TESK2
- Clonality:Monoclonal
- Clone:5G1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:10420
- Antigen synonyms:Testicular protein kinase 2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: TESK2
Clonality: Monoclonal
Clone: 5G1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human