Anti-SF3B4 Mouse Monoclonal Antibody [clone: 1B8]
Catalog # ABNOH00010262-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Splicing Factor 3b Subunit 4, 49kDa
- Antigen symbol:SF3B4
- Clonality:Monoclonal
- Clone:1B8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:10262
- Antigen synonyms:SF3B49|SF3B4|splicing factor 3b|pre-mRNA splicing factor SF3b 49 kDa subunit|49kD|spliceosomal protein|SF3b49|subunit 4|SAP49|MGC10828|spliceosome-associated protein (U2 snRNP)
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: SF3B4
Clonality: Monoclonal
Clone: 1B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human