Order Entry
Poland
ContactUsLinkComponent
Anti-POP7 Mouse Polyclonal Antibody
Anti-POP7 Mouse Polyclonal Antibody
Catalog # ABNOH00010248-B01P
Supplier:  Abnova
undefined
Anti-POP7 Mouse Polyclonal Antibody
Catalog # ABNOH00010248-B01P
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Ribonuclease P protein subunit p20
  • Antigen symbol:
    POP7
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    10248
  • Antigen synonyms:
    ribonuclease P/MRP subunit|POP7 homolog|RPP20|RPP2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pk:
    50 µg

Specifications

About this item

Type: Primary
Antigen: POP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human