Anti-SGK2 Mouse Monoclonal Antibody [clone: 7A6]
ABNOH00010110-M03
: Abnova
- Antibody type:Primary
- Antigen name:Serum/glucocorticoid regulated kinase 2
- Antigen symbol:SGK2
- Clonality:Monoclonal
- Clone:7A6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Gene ID:10110
- Antigen synonyms:Serine/threonine-protein kinase Sgk2|Serum/glucocorticoid regulated kinase 2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: SGK2
Clonality: Monoclonal
Clone: 7A6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat