Anti-DPP3 Mouse Monoclonal Antibody [clone: 3C6]
Catalog # ABNOH00010072-M07
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Dipeptidylpeptidase 3
- Antigen symbol:DPP3
- Clonality:Monoclonal
- Clone:3C6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:10072
- Antigen synonyms:DPPIII
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: DPP3
Clonality: Monoclonal
Clone: 3C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human