Anti-SH2D3A Mouse Monoclonal Antibody [clone: 3B11]
ABNOH00010045-M04
: Abnova
- Antibody type:Primary
- Antigen name:SH2 domain containing 3A
- Antigen symbol:SH2D3A
- Clonality:Monoclonal
- Clone:3B11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:10045
- Antigen synonyms:SH2 domain containing 3A|SH2D3A|NSP1|novel SH2-containing protein 1|SH2 domain-containing 3A
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: SH2D3A
Clonality: Monoclonal
Clone: 3B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human