Anti-SH2D3C Mouse Monoclonal Antibody [clone: 3B2]
ABNOH00010044-M01
: Abnova
- Antibody type:Primary
- Antigen name:SH2 domain containing 3C
- Antigen symbol:SH2D3C
- Clonality:Monoclonal
- Clone:3B2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:10044
- Antigen synonyms:NSP3|CHAT|PRO34088|SHEP1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: SH2D3C
Clonality: Monoclonal
Clone: 3B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human