Anti-NR1H4 Mouse Monoclonal Antibody [clone: 1B10]
Catalog # ABNOH00009971-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Nuclear Receptor Subfamily 1, Group H, Member 4
- Antigen symbol:NR1H4
- Clonality:Monoclonal
- Clone:1B10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:9971
- Antigen synonyms:HRR-1|BAR|FXR|RIP14|HRR1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: NR1H4
Clonality: Monoclonal
Clone: 1B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human