Order Entry
Poland
ContactUsLinkComponent
Anti-SETDB1 Mouse Monoclonal Antibody [clone: 4E8]
Anti-SETDB1 Mouse Monoclonal Antibody [clone: 4E8]
  ABNOH00009869-M22A
 :  Abnova
undefined
Anti-SETDB1 Mouse Monoclonal Antibody [clone: 4E8]
  ABNOH00009869-M22A
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    SET Domain Bifurcated 1
  • Antigen symbol:
    SETDB1
  • Clonality:
    Monoclonal
  • Clone:
    4E8
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    9869
  • Antigen synonyms:
    ESET|KMT1E|KIAA0067|KG1T|SETDB1|H3-K9-HMTase4
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    200 µl
  • Pk:
    200 µl

 

 

Type: Primary
Antigen: SETDB1
Clonality: Monoclonal
Clone: 4E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human