Anti-MTSS1 Mouse Monoclonal Antibody [clone: 4D11]
Catalog # ABNOH00009788-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Metastasis suppressor 1
- Antigen symbol:MTSS1
- Clonality:Monoclonal
- Clone:4D11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:9788
- Antigen synonyms:Metastasis suppressor YGL-1|KIAA429|MTSS1|MIM|MIMB|MIMA|Missing in metastasis protein|Metastasis suppressor protein 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: MTSS1
Clonality: Monoclonal
Clone: 4D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human