Anti-PHACTR2 Mouse Polyclonal Antibody
Catalog # ABNOH00009749-A01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Phosphatase And Actin Regulator 2
- Antigen symbol:PHACTR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:9749
- Antigen synonyms:Phosphatase and actin regulator 2 |C6orf56|KIAA0680|PHACTR2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: PHACTR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse