Anti-SOX13 Rabbit Polyclonal Antibody
ABNOH00009580-D01P
: Abnova
- Antibody type:Primary
- Antigen name:SRY (sex determining region Y)-box 13
- Antigen symbol:SOX13
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:9580
- Antigen synonyms:MGC117216|SOX 13|ICA 12|sox13|SOX13 protein|Transcription factor Sox-13|Transcription factor SOX 13|ICA12|SRY related HMG box gene 13|Islet cell antigen 12|Type 1 diabetes autoantigen|SRY Sex determining region Y-box 13|SOX13_HUMAN|SRY box 13|SRY sex determining region Y box 13|SOX 13 protein|SRY box containing gene 13|Islet cell 12|Type 1 diabetes autoantigen ICA 12|Type 1 diabetes autoantigen ICA12.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: SOX13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human