Anti-GTF2IRD1 Rabbit Polyclonal Antibody
Catalog # ABNOH00009569-D01
Dostawca: Abnova
Dane techniczne
- Antibody type:Primary
- Antigen name:GTF2I repeat domain containing 1
- Antigen symbol:GTF2IRD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Cross adsorption:No
- Gene ID:9569
- Antigen synonyms:RBAP2|hMusTRD1alpha1|WBSCR11|WBS|CREAM1|GTF3|MUSTRD1|BEN|WBSCR12
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Dane techniczne
Informacje o tym produkcie
Type: Primary
Antigen: GTF2IRD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human