Anti-HAND2 Mouse Monoclonal Antibody [clone: 4E12]
Catalog # ABNOH00009464-M04
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Heart and neural crest derivatives expressed 2
- Antigen symbol:HAND2
- Clonality:Monoclonal
- Clone:4E12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:9464
- Antigen synonyms:dHand|Hed|bHLHa26|Thing2|DHAND2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: HAND2
Clonality: Monoclonal
Clone: 4E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human