Anti-HOMER3 Mouse Polyclonal Antibody
ABNOH00009454-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Homer protein homolog 3
- Antigen symbol:HOMER3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:9454
- Antigen synonyms:VESL3|Homer3|Homer homolog 3 Drosophila|Homer 3D protein|HOMER 3|Homer neuronal immediate early gene 3|HOME3_HUMAN.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: HOMER3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human