Anti-GTF3C5 Mouse Monoclonal Antibody [clone: 3F10]
Catalog # ABNOH00009328-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:General Transcription Factor 3C, polypeptide 5
- Antigen symbol:GTF3C5
- Clonality:Monoclonal
- Clone:3F10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:9328
- Antigen synonyms:TFIIIC63|TFIIICepsilon|TFiiiC2-63
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: GTF3C5
Clonality: Monoclonal
Clone: 3F10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human