Anti-ZNHIT3 Mouse Monoclonal Antibody [clone: 2F8]
ABNOH00009326-M09
: Abnova
- Antibody type:Primary
- Antigen name:Zinc Finger, HIT-type Containing 3
- Antigen symbol:ZNHIT3
- Clonality:Monoclonal
- Clone:2F8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:9326
- Antigen synonyms:TRIP3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: ZNHIT3
Clonality: Monoclonal
Clone: 2F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human