Anti-RPS6KA5 Mouse Monoclonal Antibody [clone: 2B11]
Catalog # ABNOH00009252-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Ribosomal Protein S6 Kinase, 90kDa, Polypeptide 5
- Antigen symbol:RPS6KA5
- Clonality:Monoclonal
- Clone:2B11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:9252
- Antigen synonyms:MSK1|S6K-alpha-5|Ribosomal protein S6 kinase alpha-5|Nuclear mitogen- and stress-activated protein kinase 1|90 kDa ribosomal protein S6 kinase 5|RLPK|MSPK1|RSK-like protein kinase
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: RPS6KA5
Clonality: Monoclonal
Clone: 2B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human