Anti-SLC7A7 Mouse Monoclonal Antibody [clone: 3B10]
Catalog # ABNOH00009056-M04
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Solute Carrier Family 7 Member 7
- Antigen symbol:Solute carrier family 7, member 7
- Clonality:Monoclonal
- Clone:3B10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:9056
- Antigen synonyms:LAT3|Y+LAT1|MOP-2|y+LAT-1|LPI
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: SLC7A7
Clonality: Monoclonal
Clone: 3B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human