Anti-PSTPIP1 Mouse Monoclonal Antibody [clone: 1D5]
Catalog # ABNOH00009051-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Proline-serine-threonine Phosphatase Interacting Protein 1
- Antigen symbol:PSTPIP1
- Clonality:Monoclonal
- Clone:1D5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:9051
- Antigen synonyms:CD2BP1L|CD2BP1S|PSTPIP|PAPAS|CD2BP1|H-PIP
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PSTPIP1
Clonality: Monoclonal
Clone: 1D5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse