Anti-KAT2B Mouse Monoclonal Antibody [clone: 2B4]
Catalog # ABNOH00008850-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:K(lysine) acetyltransferase 2B
- Antigen symbol:KAT2B
- Clonality:Monoclonal
- Clone:2B4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:8850
- Antigen synonyms:CAF|P/CAF|PCAF
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: KAT2B
Clonality: Monoclonal
Clone: 2B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human