Anti-CDKL1 Mouse Monoclonal Antibody [clone: 8B2]
Catalog # ABNOH00008814-M02A
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cyclin-dependent kinase-like 1
- Antigen symbol:CDKL1
- Clonality:Monoclonal
- Clone:8B2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:8814
- Antigen synonyms:Cyclin-dependent kinase-like 1|Serine/threonine-protein kinase KKIALRE|P42
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Type: Primary
Antigen: CDKL1
Clonality: Monoclonal
Clone: 8B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human