Anti-AKR7A2 Mouse Monoclonal Antibody [clone: 2H3]
Catalog # ABNOH00008574-M15
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Aldo-keto reductase family 7 member A2
- Antigen symbol:AKR7A2
- Clonality:Monoclonal
- Clone:2H3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:8574
- Antigen synonyms:AFB1-AR 1|AKR7A2|AFAR1|AFAR|Aflatoxin B1 aldehyde reductase member 2|AKR7|Aldoketoreductase 7
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: AKR7A2
Clonality: Monoclonal
Clone: 2H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human