Anti-DEGS1 Mouse Monoclonal Antibody [clone: 3F9]
ABNOH00008560-M01A
: Abnova
- Antibody type:Primary
- Antigen name:delta(4)-desaturase, sphingolipid 1
- Antigen symbol:DEGS1
- Clonality:Monoclonal
- Clone:3F9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:8560
- Antigen synonyms:MIG15|Des-1|MLD|FADS7|DES1|DEGS-1|DEGS
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: DEGS1
Clonality: Monoclonal
Clone: 3F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human