Anti-TCAP Mouse Monoclonal Antibody [clone: 2B1]
ABNOH00008557-M01
: Abnova
- Antibody type:Primary
- Antigen name:Titin-cap
- Antigen symbol:TCAP
- Clonality:Monoclonal
- Clone:2B1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:8557
- Antigen synonyms:LGMD2G|telethonin|TELE|T-cap|CMD1N|CMH25
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: TCAP
Clonality: Monoclonal
Clone: 2B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human