Anti-UGCG Mouse Monoclonal Antibody [clone: 1E5]
Catalog # ABNOH00007357-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:UDP-glucose ceramide glucosyltransferase
- Antigen symbol:UGCG
- Clonality:Monoclonal
- Clone:1E5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:7357
- Antigen synonyms:GCS|GLCT1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: UGCG
Clonality: Monoclonal
Clone: 1E5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human