Anti-TWIST1 Mouse Monoclonal Antibody [clone: 2G12]
Catalog # ABNOH00007291-M13
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Twist-related protein 1
- Antigen symbol:TWIST1
- Clonality:Monoclonal
- Clone:2G12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:7291
- Antigen synonyms:CRS|CRS1|ACS3|SCS|bHLHa38|TWIST|BPES3|CSO|BPES2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: TWIST1
Clonality: Monoclonal
Clone: 2G12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human