Order Entry
Poland
ContactUsLinkComponent
Anti-TNNT3 Mouse Monoclonal Antibody [clone: 1F12]
Anti-TNNT3 Mouse Monoclonal Antibody [clone: 1F12]
Catalog # ABNOH00007140-M02
Supplier:  Abnova
undefined
Anti-TNNT3 Mouse Monoclonal Antibody [clone: 1F12]
Catalog # ABNOH00007140-M02
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Troponin T Type 3
  • Antigen symbol:
    TNNT3
  • Clonality:
    Monoclonal
  • Clone:
    1F12
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    7140
  • Antigen synonyms:
    beta-TnTF|troponin T fast skeletal muscle|TNTF|fast skeletal muscle troponin T|skeletal fast|troponin T type 3|fTnT|troponin-T3 skeletal fast|beta TnTF
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: TNNT3
Clonality: Monoclonal
Clone: 1F12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human