Anti-TEK Mouse Monoclonal Antibody [clone: 3A5]
Catalog # ABNOH00007010-M20
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Angiopoietin-1 Receptor
- Antigen symbol:TEK
- Clonality:Monoclonal
- Clone:3A5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:7010
- Antigen synonyms:Angiopoietin-1 receptor,EC=2.7.10.1 Tunica interna endothelial cell kinase Tyrosine-protein kinase receptor TEK Tyrosine-protein kinase receptor TIE-2 hTIE2 p140 TEK CD202b TEK TIE2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: TEK
Clonality: Monoclonal
Clone: 3A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat