Anti-SRF Mouse Monoclonal Antibody [clone: 1E1]
Catalog # ABNOH00006722-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Serum Response Factor
- Antigen symbol:SRF
- Clonality:Monoclonal
- Clone:1E1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:6722
- Antigen synonyms:Serum response Factor (c-fos Serum Response Element-binding transcription Factor|MCM1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: SRF
Clonality: Monoclonal
Clone: 1E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human