Order Entry
Poland
ContactUsLinkComponent
 
Anti-SPP1 Mouse Monoclonal Antibody [clone: 4D8]
Catalog # ABNOH00006696-M18
Supplier:  Abnova
undefined
Anti-SPP1 Mouse Monoclonal Antibody [clone: 4D8]
Catalog # ABNOH00006696-M18
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Secreted Phosphoprotein 1
  • Antigen symbol:
    SPP1
  • Clonality:
    Monoclonal
  • Clone:
    4D8
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    6696
  • Antigen synonyms:
    ETA-1|Opnl|Bsp|OP|2AR|BNSP|BSPI|Eta|Ric|Apl-1|Opn|Spp-1|Osteopontin
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: SPP1
Clonality: Monoclonal
Clone: 4D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human