Order Entry
Poland
ContactUsLinkComponent
Anti-SLC7A1 Mouse Monoclonal Antibody [clone: 2B9]
Anti-SLC7A1 Mouse Monoclonal Antibody [clone: 2B9]
Catalog # ABNOH00006541-M02
Supplier:  Abnova
undefined
Anti-SLC7A1 Mouse Monoclonal Antibody [clone: 2B9]
Catalog # ABNOH00006541-M02
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Solute Carrier Family 7 (cationic amino acid transporter y+ system) Member 1
  • Antigen symbol:
    Solute carrier family 7, member 1
  • Clonality:
    Monoclonal
  • Clone:
    2B9
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    6541
  • Antigen synonyms:
    ATRC1|REC1L|HCAT1|CAT-1|ERR
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: SLC7A1
Clonality: Monoclonal
Clone: 2B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human