Anti-MAPK12 Mouse Monoclonal Antibody [clone: 2C1]
ABNOH00006300-M04
: Abnova
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase 12
- Antigen symbol:MAPK12
- Clonality:Monoclonal
- Clone:2C1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:6300
- Antigen synonyms:P38GAMMA|PRKM12|MAPK 12|ERK3|SAPK-3|ERK6|SAPK3|ERK-6
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: MAPK12
Clonality: Monoclonal
Clone: 2C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human