Anti-S100 A7 Mouse Monoclonal Antibody [clone: 1C5-C6]
Catalog # ABNOH00006278-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:S100 calcium binding protein A7
- Antigen symbol:S100A7
- Clonality:Monoclonal
- Clone:1C5-C6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:6278
- Antigen synonyms:S100A7c|PSOR1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: S100 A7
Clonality: Monoclonal
Clone: 1C5-C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human