Anti-RXRA Mouse Monoclonal Antibody [clone: 3F5]
Catalog # ABNOH00006256-M34J
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Retinoid X Receptor, Alpha
- Antigen symbol:RXRA
- Clonality:Monoclonal
- Clone:3F5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:6256
- Antigen synonyms:ribonucleotide reductase M1 Polypeptide|NR2B1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: RXRA
Clonality: Monoclonal
Clone: 3F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human