Anti-PTPRN Mouse Polyclonal Antibody
Catalog # ABNOH00005798-B01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Protein tyrosine phosphatase receptor type N
- Antigen symbol:PTPRN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5798
- Antigen synonyms:ICA512|IA-2|IA2|IA-2/PTP|FLJ16131|R-PTP-N
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: PTPRN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human