Anti-PTGIS Mouse Polyclonal Antibody
Catalog # ABNOH00005740-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Prostaglandin I2 Synthase
- Antigen symbol:PTGIS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5740
- Antigen synonyms:PTGI|CYP8A1|PGIS|CYP8
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: PTGIS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human