Anti-PROX1 Mouse Monoclonal Antibody [clone: 1E7]
Catalog # ABNOH00005629-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Prospero Homeobox 1
- Antigen symbol:PROX1
- Clonality:Monoclonal
- Clone:1E7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5629
- Antigen synonyms:PROX-1|prospero-related homeobox 1|prospero homeobox protein 1|homeobox prospero-like protein PROX1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PROX1
Clonality: Monoclonal
Clone: 1E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human