Order Entry
Poland
ContactUsLinkComponent
Anti-MAP2K5 Mouse Monoclonal Antibody [clone: 1E5]
Anti-MAP2K5 Mouse Monoclonal Antibody [clone: 1E5]
Catalog # ABNOH00005607-M03
Supplier:  Abnova
undefined
Anti-MAP2K5 Mouse Monoclonal Antibody [clone: 1E5]
Catalog # ABNOH00005607-M03
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Mitogen-activated protein kinase Kinase 5
  • Antigen symbol:
    MAP2K5
  • Clonality:
    Monoclonal
  • Clone:
    1E5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    5607
  • Antigen synonyms:
    PRKMK5|MEK5|MAPKK5|HsT17454
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: MAP2K5
Clonality: Monoclonal
Clone: 1E5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human